missing translation for 'onlineSavingsMsg'
Learn More

Morpholine, 99+%, Pure, Fisher Chemical

Product Code. 10498600
Click to view available options
Quantity:
500 mL
Packaging:
Plastic Coated Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 110-91-8
Melting Point -5°C
Color Colorless
Boiling Point 126°C
Packaging Plastic Coated Glass bottle
Molecular Formula C4H9NO
MDL Number MFCD00005972
Quantity 500 mL
Synonym 1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro
InChI Key YNAVUWVOSKDBBP-UHFFFAOYSA-N
SMILES C1COCCN1
IUPAC Name morpholine
Molecular Weight (g/mol) 87.12
PubChem CID 8083
Vapor Density 3
ChEBI CHEBI:34856
Viscosity 2.23cp at 20°C
Formula Weight 87.12g/mol
Vapor Pressure 11mbar at 20°C
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Morpholine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.