missing translation for 'onlineSavingsMsg'
Learn More

Motor Octane Number Standard ASTM D 2700 (Nominal value: 86.9), Paragon Scientific™

Fuels Testing Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Octane Number (Motor Method) ASTM D2700

Brand:  Paragon Scientific CRM-OMGA


Product Code. 16000192

  • 754.00 € / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Manufactured in accordance with ASTM / EN or IP test methods
  • Certified by Round Robin Method by independant test laboratories
  • Ensures good interlaboratory correlation
  • Supplied in tamper evident UN compliant security pack
  • Pack size 1000mL
  • 12 months shelf life
Specifications

Specifications

UN 1203
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 1B
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361-Suspected of damaging fertility or the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
  • P391-Collect spillage.
  • P403+P235-Store in a well-ventilated place. Keep cool.

Supplemental information
  • MIXTURE LIST-Contains : Gasoline : low boiling point naphtha

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Nominal values quoted. Certified Values will vary from batch to batch.