missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Moulded Polyethylene Connecting Hook

Product Code. 11791296
missing translation for 'orderingAttributeHoverText'
Color:
Black
Yellow
missing translation for 'unitSize'
1 pack
This item is not returnable. View return policy

Product Code. 11791296

missing translation for 'mfr': Brady™ 078259

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Used to attach lengths of chain, our durable B-900 Moulded Polyethylene Connecting Hooks are available to match our posts and chain.

  • Used to attach lengths of chain, our durable B-900 polyethylene Connecting Hooks are available to match our posts and chain.

Specifications

Quantity 36 Pk.
Type Hook
Color Black
Material Polyethylene
For Use With (Application) Traffic Control
Width (English) 2 in.
Width (Metric) 50.8 mm
Label Type Stanchion Accessories
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.