Learn More
Invitrogen™ Mouse ADAMTS4 (aa 394-403) Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP234
Product Code. 11544880
  • 159.00 € / 50µg
Explore more special offers
Description
Description
Mouse ADAMTS4 (residues 394-403) Synthetic Peptide, PEP-234, from Invitrogen.
PEP-234 is a 11 amino acid synthetic peptide corresponding to amino acid residues 394-403 from mouse ADAMTS4. The sequence of this peptide is (amino to carboxy terminus): C V (394) M A H V D P E E P (403).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA1-1749A. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
240913
purified
Mouse ADAMTS4 (aa 394-403)
-20°C. Avoid freeze-thaw cycles.
Lyophilized
Control
Yes
Peptide
RUO - research use only
Mouse
Q8BNJ2
0.5 mg/mL
Synthetic Peptide
peptide with no preservative
Adamts4
No preservative
50 μg
C V(394) M A H V D P E E P(403)