missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse anti-Human IgE Secondary Antibody
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen™ SA119260
This item is not returnable.
View return policy
Description
This antibody recognizes an epitope different from the ones recognized by BE5 and 4H10 antibodies to IgE. The epitope is located within the amino acids 103-115 (WSDYNFDYSSSEE).
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Human IgE | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| Purified human IgE. | |
| 100 μg | |
| Secondary | |
| 4°C, do not freeze | |
| Liquid |
| ELISA, Flow Cytometry | |
| 4G7.325 | |
| Unconjugated | |
| Mouse | |
| Sequential chromatography | |
| RUO | |
| Human | |
| Secondary Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction