Learn More
Invitrogen™ Mouse anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP
Mouse Polyclonal Secondary Antibody
Brand:  Invitrogen™ 31420
Product Code. 10529673
  • 273.00 € / 1.50mL
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31420 has been successfully used in Western blot, and ICC applications. Antibody Specificity: Based on immunoelectrophoresis, the antibody reacts with the heavy chains on human IgG and with light chains common to most human immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. This antibody exhibits minimal inherent cross-reactivity to mouse serum proteins. It has been passed through solid-phase immunoadsorbent gel to minimize any actual cross-reaction with mouse serum proteins. However, these antibodies may cross-react with immunoglobulins from other species. Restoration and Storage: Store product at 4°C until opened. Restore with 1.5 mL distilled water (0.8 mg/mL after restoration). Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product may be stored for several weeks at 4°C as undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Human IgG (H+L) Cross-Adsorbed
Polyclonal
HRP
Mouse
1.5 mL
Secondary
4°C
Lyophilized
Immunohistochemistry, Western Blot, Immunocytochemistry
0.8 mg/mL
PBS with 15mg/mL BSA and no preservative; pH 7.6
Antigen affinity chromatography
RUO
Human
Secondary Antibody