missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Human IgG1 Fc Secondary Antibody, HRP

Mouse Monoclonal Secondary Antibody

Brand:  Invitrogen™ A10648

219.45 EUR valid until 2025-12-31
BEST PRICE on Promo! Use promo code "24159" to get your promotional price.



Product Code. 10070738

  • 231.00 € / 200µg
Spotlight
Use promo code 24159
View all promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Storage and reconstitution: when stored desiccated at or below -20°C, the lyophilized powder is stable for at least one year. Reconstitute using 200 μL PBS, pH 7.2, to yield a 1 mg/mL stock solution containing 0.02% thimerosal. After reconstitution, store product at 4°C. For prolonged storage after reconstitution, add glycerol to a final concentration of 50% (v/v), aliquot, and store at or below -20°C. When stored properly, the solution is stable for approximately three months.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Human IgG1 Fc
Monoclonal
HRP
Mouse
Purified
RUO
Human
Secondary Antibody
IgG1 κ
ELISA, Immunohistochemistry, Western Blot
HP6069
PBS with 0.02% thimerosal
IgG gamma 1.
200 μg
Secondary
-20°C
Lyophilized
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Mouse anti-Human IgG1 Fc Secondary Antibody, HRP

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : thimerosal

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.