Learn More
Mouse anti-Human Kappa Light Chain Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand:  Invitrogen SA119155
Product Code. 11545020
  • 272.00 € / 100µg
Explore more special offers
Description
Description
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
MEM-09
Unconjugated
Mouse
100 μg
Secondary
4° C, do not freeze
IgG1
Monoclonal
1 mg/mL
PBS with 15mM sodium azide; pH 7.4
Crude thymus membrane fraction.