missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human Lambda Light Chain Secondary Antibody, FITC, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MH10611
This item is not returnable.
View return policy
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility. This antibody recognizes free light chains as well as those that are bound to heavy chains.
Human Lambda Light Chain Monoclonal antibody specifically detects Human Lambda Light Chain in Human samples. It is validated for Flow Cytometry.Specifications
Human Lambda Light Chain, Mouse anti-Human Lambda Light Chain Secondary Antibody, FITC | |
Monoclonal | |
FITC | |
Mouse | |
500 μL | |
Secondary | |
4° C, store in dark | |
IgG2a |
Flow Cytometry | |
HP6054 | |
PBS with BSA, sucrose and 0.1% sodium azide | |
purified | |
ASR | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction