Learn More
Mouse anti-Rat IgG1 (Heavy chain) Secondary Antibody, Biotin, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand:  Invitrogen MA516815
Product Code. 15256059
  • 656.00 € / 500µg
Explore more special offers
Description
Description
Mouse anti rat IgG1, clone MARG1-2 recognizes the heavy chain of the rat immunoglobulin G1 subclass. No cross-reactivity has been found with other rat immunoglobulin classes or subclasses.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rat IgG1 (Heavy chain)
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide
Rat IR27 IgG1 myeloma protein
500 μg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG1
ELISA
MARG1-2
Biotin
Mouse
Affinity chromatography
RUO
Rat