missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse (CD-1) Cryopreserved Hepatocytes, Female

Mouse (CD-1) Cryopreserved Hepatocytes, Female

Brand:  Gibco™ MSCS20

Product Code. 10819040

  • 730.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

These cryopreserved female mouse (CD-1) hepatocytes have been characterized for general Phase I and Phase II drug metabolizing enzyme activities.

Cells per vial: 4-8 million

Get the Results You Need with GIBCO Hepatocytes
High quality hepatocytes, with high viabilities, in vivo-like enzyme expression levels, and proper cell morphology increase the ability to draw in vitro⁄in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure you receive the highest quality cryopreserved hepatocytes.

Each lot is tested for:
  • Phase I and II enzyme activities
  • Proper morphology
  • Viabilities generally >75%

    For research use only. Not intended for human or animal therapeutic or diagnostic use.

    Related Links:

    Learn more about our cryopreserved hepatocytes

    Learn about our full line of hepatocytes ,

    Note:
    These products require shipment in LN2 vapor dewars therefore additional fees may apply.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Vials of cryopreserved hepatocytes should be stored in a cryostorage dewar or -135°C freezer.
    Hepatocyte Cell Culture
    Female
    Pooled
    Gibco™
    1 vial
    Hepatocytes
    Cryopreserved
    Mouse
    4-8 x 106
    Hepatocytes
    Phase I and II Enzyme Activity, Proper Morphology, Viability Generally >75%
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.