missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse FGF-10 Recombinant Protein, PeproTech®

Recombinant Protein

83.75 € - 3100.00 €

Specifications

Accession Number O35565
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 14165
Molecular Weight (g/mol) 17 kDa
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17843863
View Documents
Gibco™
450-61-100UG
100 μg
622.00 €
100µg
Please to purchase this item. Need a web account? Register with us today!
17844793
View Documents
Gibco™
450-61-1MG
1 mg
3100.00 €
1mg
Please to purchase this item. Need a web account? Register with us today!
17893923
View Documents
Gibco™
450-61-250UG
250 μg
1088.00 €
250µg
Please to purchase this item. Need a web account? Register with us today!
17804073
View Documents
Gibco™
450-61-25UG
25 μg
203.00 €
25µg
Please to purchase this item. Need a web account? Register with us today!
17894363
View Documents
Gibco™
450-61-500UG
500 μg
1854.00 €
500µg
Please to purchase this item. Need a web account? Register with us today!
17804583
View Documents
Gibco™
450-61-5UG
5 μg
83.75 €
5µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Recombinant Murine FGF-10 is a 17.0 kDa protein consisting of 148 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

FGF10 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein exhibits mitogenic activity for keratinizing epidermal cells, but essentially no activity for fibroblasts, which is similar to the biological activity of FGF7. Studies of the mouse homolog of suggested that this gene is required for embryonic epidermal morphogenesis including brain development, lung morphogenesis, and initiation of lim bud formation. This protein is also implicated to be a primary factor in the process of wound healing.
Specifications

Specifications

O35565
protein with no preservative
17 kDa
E. coli
<1 EU/ μg
FGF10
Determined by dose-dependent ability to reduce tetrazolium salt, WST-8, by dehydrogenase activities of BaF3 cells expressing FGF receptors using Cell Counting Kit-8 (CCK-8).
Recombinant
-20°C
Lyophilized
Functional Assay
14165
Mouse FGF-10
RUO
AEY17; BB213776; FGF; Fgf10; FGF-10; Fibroblast growth factor; Fibroblast growth factor 10; Gsfaey17; Keratinocyte growth factor 2; produced by fibroblasts of urinary bladder lamina propria
FGF10
Unconjugated
SSAGRHVRSY NHLQGDVRWR RLFSFTKYFL TIEKNGKVSG TKNEDCPYSV LEITSVEIGV VAVKAINSNY YLAMNKKGKL YGSKEFNNDC KLKERIEENG YNTYASFNWQ HNGRQMYVAL NGKGAPRRGQ KTRRKNTSAH FLPMTIQT
E. coli
≥ 95% by SDS-PAGE gel and HPLC analyses.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.