missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse FGF-4 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
80.50 € - 2980.00 €
Specifications
Accession Number | P11403 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 14175 |
Molecular Weight (g/mol) | 19 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17834363
|
Gibco™
450-57-100UG |
100 μg |
598.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17804573
|
Gibco™
450-57-1MG |
2 x 500 μg | N/A | ||||||
17853933
|
Gibco™
450-57-250UG |
250 μg |
1044.00 €
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17843853
|
Gibco™
450-57-25UG |
25 μg |
195.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17804793
|
Gibco™
450-57-500UG |
500 μg |
1784.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17873853
|
Gibco™
450-57-5UG |
5 μg |
80.50 €
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibroblast growth factor 4 (FGF4) is a member of the fibroblast growth factor (FGF) family that possess broad mitogenic and cell survival activities and play key roles in growth and survival of stem cells during embryogenesis, tissue regeneration, and carcinogenesis. FGF4 was identified by its strong oncogenic transforming activity and is a potent angiogenic factor, expressed in several highly vascularized tumors and also in adult mouse testis, intestine, and brain. Studies on the mouse homolog suggests a function in bone morphogenesis and limb development through the sonic hedgehog (SHH) signaling pathway. Furthermore, FGF4 regulates neural progenitor cell proliferation and neuronal differentiation. Recent studies show a growth-promoting role for FGF4 in human embryonic stem cells and a putative feedback inhibition mechanism by a novel FGF4 splice isoform that may serve to promote differentiation at a later stages of development.Specifications
P11403 | |
protein with no preservative | |
19 | |
E. coli | |
RUO | |
FGF; FGF4; Fgf-4; Fgfk; Fibroblast growth factor; fibroblast growth factor 4; fibroblast growth factor 4 splice isoform; HBGF-4; heparin secretory transforming protein 1; heparin secretory-transforming protein 1; heparin-binding growth factor 1; heparin-binding growth factor 4; HST; Hst1; hst-1; HSTF1; HSTF-1; human stomach cancer, transforming factor from FGF-related oncogene; kaposi sarcoma oncogene; Kfgf; K-FGF; K-fibroblast growth factor; KS3; oncogene HST; transforming protein KS3 | |
Mouse | |
Recombinant | |
Lyophilized |
Functional Assay | |
14175 | |
Mouse FGF-4 | |
-20°C | |
<1 EU/μg | |
Fgf4 | |
Unconjugated | |
APNGTRHAEL GHGWDGLVAR SLARLPVAAQ PPQAAVRSGA GDYLLGLKRL RRLYCNVGIG FHLQVLPDGR IGGVHADTRD SLLELSPVQR GVVSIFGVAS RFFVAMSSRG KLFGVPFFTD ECKFKEILLP NNYNAYESYA YPGMFMALSK NGRTKKGNRV SPTMKVTHFL PRL | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |