Learn More
Mouse IL-10 ELISPOT Pair, BD
Convenient set of unlabelled anti-IL-10 capture antibody and a biotinylated anti-IL-10 detection antibody
Brand: BD Biosciences 557982
758.84 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes: Mouse IL-10 ELISPOT Capture Antibody (no azide/low endotoxin), Mouse IL-10 ELISPOT Detection Antibody (biotinylated) and a Certificate of Analysis that provides lot-specific optimal reagent concentrations
Description
Specifications
5 x 96-well Plates |
Product Content Correction
Product Title
Mouse IL-10 ELISPOT Pair, BD > For 5 x 96-well plates
Spot an opportunity for improvement?Share a Content Correction