Learn More
Mouse IL-2 ELISPOT Pair, BD
Convenient set of unlabelled anti-IL-2 capture antibody and a biotinylated anti-IL-2 detection antibody
Brand: BD Biosciences 551876
758.84 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes: Mouse IL-2 ELISPOT Capture Antibody (no azide/low endotoxin), Mouse IL-2 ELISPOT Detection Antibody (biotinylated) and a Certificate of Analysis that provides lot-specific optimal reagent concentrations
Description
Specifications
5 x 96-well Plates |