Learn More
Mouse IL-4 ELISPOT Pair, BD
Convenient set of unlabelled anti-IL-4 capture antibody and a biotinylated anti-IL-4 detection antibody
Brand:  BD Biosciences 551878
758.84 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15866718
  • 760.00 € / Each
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Includes: Mouse IL-4 ELISPOT Capture Antibody (no azide/low endotoxin), Mouse IL-4 ELISPOT Detection Antibody (biotinylated) and a Certificate of Analysis that provides lot-specific optimal reagent concentrations