missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse LIGHT Recombinant Protein, PeproTech®

E. coli Recombinant Protein

80.50 € - 2980.00 €

Specifications

Accession Number Q9QYH9
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 50930
Molecular Weight (g/mol) 20.1
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17802963
View Documents
Gibco™
315-12-100UG
100 μg
598.00 €
100µg
Please to purchase this item. Need a web account? Register with us today!
17812583
View Documents
Gibco™
315-12-1MG
1 mg
2980.00 €
1mg
Please to purchase this item. Need a web account? Register with us today!
17882363
View Documents
Gibco™
315-12-20UG
20 μg
195.00 €
20µg
Please to purchase this item. Need a web account? Register with us today!
17812373
View Documents
Gibco™
315-12-250UG
250 μg
1044.00 €
250µg
Please to purchase this item. Need a web account? Register with us today!
17802583
View Documents
Gibco™
315-12-500UG
500 μg
1784.00 €
500µg
Please to purchase this item. Need a web account? Register with us today!
17802373
View Documents
Gibco™
315-12-5UG
5 μg
80.50 €
5µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.
Specifications

Specifications

Q9QYH9
protein with no preservative
20.1
E. coli
RUO
CD258; Herpes virus entry mediator ligand; herpesvirus entry mediator ligand; HVEML; HVEM-L; Light; LTg; Ly113; sCD258; sLIGHT; soluble CD258; soluble LIGHT; Tnfsf14; TR2; tumor necrosis factor (ligand) superfamily, member 14; tumor necrosis factor ligand 1 D; tumor necrosis factor ligand superfamily member 14; Tumor necrosis factor ligand superfamily member 14, membrane form; Tumor necrosis factor ligand superfamily member 14, soluble form; tumor necrosis factor superfamily member 14; UNQ391/PRO726
Mouse
Recombinant
Lyophilized
ELISA, Functional Assay, Western Blot
50930
Mouse LIGHT
-20°C
<1 EU/μg
TNFSF14
Unconjugated
MRLHQRLGDI VAHLPDGGKG SWEKLIQDQR SHQANPAAHL TGANASLIGI GGPLLWETRL GLAFLRGLTY HDGALVTMEP GYYYVYSKVQ LSGVGCPQGL ANGLPITHGL YKRTSRYPKE LELLVSRRSP CGRANSSRVW WDSSFLGGVV HLEAGEEVVV RVPGNRLVRP RDGTRSYFGA FMV
≥ 98% by SDS-PAGE gel and HPLC analyses.
Videos
SDS
Documents

Documents

Product Certifications