missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse MMP-1a (NP_114395) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Specifications
Specifications
| Gene | MMP-1 |
| Accession Number | NP_114395 |
| Gene ID (Entrez) | 4312 |
| Gene Alias | CLGmatrix metalloprotease 1, CLGN, EC 3.4.24, EC 3.4.24.7, Fibroblast collagenase, interstitial collagenase, matrix metallopeptidase 1 (interstitial collagenase), matrix metalloproteinase 1 (interstitial collagenase), Matrix metalloproteinase-1, MMP1, MMP-1 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction