missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse NGF 7S Subunit Native Protein
Mouse NGF 7S Subunit Native Protein
Brand: Gibco™ 13290010
This item is not returnable.
View return policy
Description
Mouse NGF 7S Subunit Native Protein for Western Blot, IHC, ELISA, Ctrl, FN
NGF 7S, Murine, Natural, is a 130 to 140kDa complex that regulates the development and maintenance of sympathetic and some sensory neurons NGF is used in studies of nerve cell growth and differentiation. It can also be used as a medium supplement to support the growth or survival of neuronal cells in culture.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; natural Mouse Nerve Growth Factor 7S (NGF 7S) specific activity is determined by measuring neurite outgrowth in chick embryo dorsal root ganglia
- Proven Compatibility: Gibco proteins are bioassayed with Gibco media
Shipping Condition: Dry Ice.
Specifications
Cell Culture | |
Mouse | |
100 μg | |
Mammalian | |
Native, Ligand | |
Neurotrophic Factors |
Nerve Growth Factor 7S (NGF 7S) | |
Store in freezer (-5 to -30°C). | |
Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Neurobiology, Signal Transduction, Oncology, Stem Cell Research, Inflammation, Differentiation, Cardiovascular Research |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.