missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse Olfactomedin-1 ELISA Kit
Mouse Olfactomedin-1 quantitates mouse Olfactomedin-1 in serum, plasma, supernatant.
Brand: Invitrogen™ EM58RB
This item is not returnable.
View return policy
Description
Mouse Olfactomedin-1 quantitates mouse Olfactomedin-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse Olfactomedin-1.
The Olfactomedin family comprises a diverse group of secreted glycoproteins, which includes OLFM1 (Noelin-1), OLFM2 (Noelin-2), OLFM3 (Noelin-3), OLFM4 (Noelin-4), tiarin, pancortin, gliomedin and mycocilin. These proteins are implicated in the development of the nervous system. Specifically, OLFM1 and OLFM2 expression is observed in the neural plate and neural crest, as well as in the cranial ganglia in mouse at E8-10, and later in brain tissue and in the zone of polarizing activity in the limb. Overexpression of OLFM1 causes an excess of neural crest emigrations and prolonged neural crest production. OLFM2 participates in the regulation of the development of the anterior nervous system. An Arg144Gln mutation in OLFM2 has been implicated as a possible cause for open-angle glaucoma (OAG).Specifications
1B426B; AMY; AW742568; D2Sut1e; neuroblastoma protein; neuronal olfactomedin-related ER localized protein; Noe1; Noel; NOEL1; noelin; Noelin 1; Noelin 2; NOELIN1; OlfA; olfactomedin 1; olfactomedin related ER localized protein; olfactomedin-1; OLFM1; pancortin; pancortin 1; Pancortin 1-4; Pancortin3 | |
0.084 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
OLFM1 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
0.084 to 20 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
O88998 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Serum, 50 μL; Plasma, 50 μL; Supernatant, 50 μL | |
OLFM1 | |
4 hr 45 min |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction