Learn More
R&D Systems™ Mouse Tyro3/Dtk DuoSet ELISA
Sufficient for at least 15 96-well plates
Brand:  R&D Systems™ DY759
757.60 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
Product Code. 30021110
  • 947.00 € / 1 set
Explore more special offers
Description
Description
The Mouse Tyro3/Dtk DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse Dtk/TYRO3.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Biotin
ELISA
Brt, BYK, Dtk, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY
125.00 to 8000 pg/mL
22174
Store the unopened product at 2°C to 8°C. Do not use past expiration date.