Learn More
R&D Systems™ Mouse XL Cytokine Luminex™ Performance Premixed Kit
Brand:  R&D Systems™ FCSTM20-28
 View more versions of this product
Product Code. 18743937
  • 6250.00 € / Each
Explore more special offers
Specifications
28 Plex
Mouse
Store the unopened product at 2°C to 8°C. Do not use past expiration date.