missing translation for 'onlineSavingsMsg'
Learn More

Welch™ MP 301 V Vacuum Pump

Proven diaphragm technology offers a double benefit to the user

Brand:  Welch™ 411744

Product Code. 11842243

  • 3688.80 € / Each
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Low weight and excellent ergonomics
  • Proven diaphragm technology offers a double benefit to the user
  • ON/OFF switch and internal protective thermal switch for the motor, mains cable and plug
  • Carry handle
  • Vibration isolating feet
  • MP models come with and exhaust silencer
  • MPC models come with a gas ballast valve
  • analytically pure, oil free vacuum
  • User friendly
  • Light weight, rugged design
  • Designed for permanent operation
  • Maintenance-free drive system and proven long diaphragm life
  • Minimal operation vibration
  • Wide vacuum and flow range to match application
  • Superior chemical resistance (MPC models)
Specifications

Specifications

38/41 L/min.
380 mm
370 w
38/41 L/min.
170 mm
Vacuum Pump
230 V
230 V
MP 301 V
18.3 kg
50/60 Hz
230 mm
<1 mbar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.