missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 1/4 in. Ceramic Spheres

Use to break hard/brittle samples. Specialty applications for the 1/4 in. Ceramic Spheres include RNA extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.

82.70 € - 1160.00 €

Specifications

Bead Type Zirconium oxide coated ceramic
Content And Storage Room Temperature
Sample Type Bone, Heart, Lung, Seeds, Skeletal Muscle, Spores
For Use With (Application) Break hard/brittle samples
Product Type Bulk Matrix
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15582384
View Documents
MP Biomedicals™
116540034
50 Pk.
82.70 €
Pack of 50
Please to purchase this item. Need a web account? Register with us today!
11432400
View Documents
MP Biomedicals™
116540422
1000 Pk.
1160.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
10548285
View Documents
MP Biomedicals™
116540424
100 Pk.
184.00 €
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
11412400
View Documents
MP Biomedicals™
116540412
500 Pk.
652.00 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Zirconium Oxide-coated ceramic grinding spheres
  • Chemically inert
  • Component of Lysing Matrix A and M tubes
Specifications

Specifications

Zirconium oxide coated ceramic
Bone, Heart, Lung, Seeds, Skeletal Muscle, Spores
Bulk Matrix
1/4 in. Zirconium Oxide Banded Satellite for Grinding
Room Temperature
Break hard/brittle samples
Unlimited
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ 1/4 in. Ceramic Spheres

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.