Learn More
MP Biomedicals™ Basal Medium Eagle Modified Vitamin Solution 100X Without L-Glutamine
Basal Medium Eagle Modified Vitamin Solution 100X Without L-Glutamine is for use with Mammalian Cell Culture to provide optimum cell growth.
Brand:  MP Biomedicals™ 091600449
 View more versions of this product
Product Code. 17367469
  • 46.85 € / 100mL
Explore more special offers
Description
Description
  • Chemically-defined essential components
  • Lot-to-lot consistency
  • Animal-component free media
  • No proteins, hormones, or other growth factors
  • No biological contamination such as viruses, mycoplasma, or prions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 X
Vitamin Supplement
No Glutamine
100 mL