missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Binding Matrix
Binding Matrix is a silica slurry that binds DNA from lysates for further purification.
Brand: MP Biomedicals™ 116540408
188.93 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- It is used to extract genomic DNA from plants, animals, bacteria, yeast, algae, and fungi, soil and environmental samples.
- Binding Matrix is a component included in the FastDNA and all FastDNA Spin Kits and is used to bind DNA from a lysate.
- Once bound, impurities and inhibitors are washed away with the SEWS-M solution.
- The resulting pure DNA is then eluted from the Binding Matrix with either an elution solution or DDI water.
Specifications
Seeds, Spores, Bacteria (Gram +/-), Animal Tissues, Feces, Food, Fungi, Plants, Roots, Soil, Wastewater, Yeast | |
Sample Preparation, Sample Lysis, Nucleic Acid Isolation and Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation | |
2 x 30 mL |
FastPrep-24, FastPrep 120, FastPrep-96, SuperFastPrep-1 | |
Binding Matrix | |
Up to 500 mg Per Prep |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Binding Matrix
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : guanidine thiocyanate
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.