missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Bovine Fibrinogen
Suitable as a substrate for cell culture and in clotting assays
Brand: MP Biomedicals™ 08820225
This item is not returnable.
View return policy
Description
Prepared by extensive washing of cryoprecipitate followed by a buffer extraction to remove unwanted proteins and precipitation with ammonium sulfate. Lyophilized powder. Clottable protein >90%.
Physical Characteristics
- Appearance: Off-white powder
- Clottable Protein (anhydrous): ≥90%
- Moisture: ≤5%
- Reported: Total protein, pH
Specifications
Mammalian Cell Culture | |
6.4 | |
Bovine Plasma | |
Lyophilized |
Bovine Fibrinogen 90% Clottable from Bovine Plasma | |
100g | |
<-10°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction