missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Bovine Transferrin, (HOLO)
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
500 mg
missing translation for 'unitSize'
100mg
500mg
Descripción
Transferrin is a 77kDa plasma glycoprotein with the capacity to bind ferric ions and as such is a source of bio-available iron. The product is frequently used as a nutrient in cell and microbial culture and as a media supplement in the production of bio-pharmaceutical products. Transferrin has also been shown to function as a factor in the growth stimulation of many cell types, such as granulocytes, macrophages and endothelial cells. Chromatographically purified.
Physical characteristics:
- Appearance: Reddish-brown powder
- Purity: ≥95% w/w
- Iron: ≥1200 mg/g protein
- Moisture: ≤5%
- pH (3%): 6.0 to 8.0
- Solubility: Dissolves in <20 min.
- Bioburden: ≤100cfu/g
- Endotoxin: ≤1 EU/mg
- Mycoplasma: Not detected
- Virus: Not detected
Especificaciones
Especificaciones
| pH Range | 6 to 8 |
| Quantity | 500 mg |
| Source | Bovine Serum |
| Storage Requirements | 4°C |
| Product Type | Bovine Transferrin (HOLO) |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido