missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Corn Syrup
Dual acid-enzyme conversion syrup
Brand: MP Biomedicals™ 0210141301
93.46 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Contains three times the maltose of regular corn syrup.
- Liquid
- Maltose content approximately 30%
- Processed using acid only
Specifications
Animal Diet | |
Corn syrup are used in commercially prepared foods as a thickener, a sweetener and as a humectant - an ingredient that retains moisture and thus maintains a food's freshness | |
15°C to 30°C |
Liquid | |
1 gal. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction