missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ CSM-HIS-LEU, Powdered Media

Dropout of Complete Supplement Mixture (CSM) of amino acids for S. cereviseae growth media

Brand:  MP Biomedicals™ 114520412

 View more versions of this product

Product Code. 11317919

  • 92.45 € / 10g
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Formulation

  • Add 0.67g of CSM-His-Leu to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined double drop-out medium

Applications:

Yeast cell culture, Gene expression, Transcription regulation

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Molecular biology Certified™
Powder
Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 0mg/L, L-Isoleucine 50mg/L, L-Leucine 0mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, L-Tyrosine 50mg/L, Uracil 20mg/L, L-Valine 140mg/L
10 g
15°C to 30°C
Yeast Media
For growth of Saccharomyces cerevisiae
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.