missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Dextrose

Monohydrate

Brand:  MP Biomedicals™ 0290559425

167.24 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11422781

  • 181.00 € / 25lb

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Dextrose monohydrate is a common natural sugar involved in processes such as energy production, glycosylation and formation of glycans that provide structure to cells. It is involved in a detrimental process in cells called glycation. It is used as a supplement for cell culture and in numerous cellular processes.

Formula: C6H12O6•H2O

Formula Weight: 198.18

CAS No.: 14431-43-7

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Animal Diet
Used as a supplement for cell culture and in numerous cellular processes
15°C to 30°C
Powder
11.33 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers