Learn More
MP Biomedicals™ DNA-Erase™ Decontaminant
Removes DNA contamination
Brand:  MP Biomedicals™ 04821805
84.64 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11485802
  • 133.00 € / 500mL
Edge
Explore more special offers
Description
Description
DNA-Erase™ is a non-alkaline, non-corrosive and non-carcinogenic cleansing solution that is highly active against DNA contamination. Presence of contaminant DNA at a PCR work area can result in DNA amplification artifacts. The solution contains a surfactant plus an agent that destroys DNA. By following the simple decontamination instructions, DNA is completely destroyed and removed.
  • Eliminates PCR artifacts from all laboratory surfaces and apparatus, including pipetters
  • Stable, heat-resistant, and noncorrosive
  • Supplied as ready-to-use formula
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Decontaminant Solution
16.9 oz.
500 mL
Removal of DNA contamination
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals™ DNA-Erase™ Decontaminant > 500mL