missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ EcoChrome™ Silica TLC Plate

Used as a defoamer component. In hydrated form, it is used in toothpaste as a hard abrasive to remove tooth plaque. SiO

Brand:  MP Biomedicals™ 0204621

194.35 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.


Product Code. 11313599

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

EcoChrom™ Silica Plates, cover a wide range of Thin-Layer Chromatography applications. Their uniform surface makes MP silica plates suitable for quantitative densitometric UV/VIS evaluation. They are available without any fluorescent indicator or with fluorescent indicators for 254 nm or for 254 and 366 nm activation wavelength.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

60 Å
Silica TLC Plate
Quantitative Densitometric UV/VIS Evaluation
0.5 mm
20 cm
20/Pk.
254 nm
Silica Gel
20 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.