missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ CoolPrep™ Adapter for 24 x 2 mL Tube Holder on FastPrep-24

CoolPrep™ Adapter for FastPrep-24 instrument is a cryogenic adapter that allows simultaneous lysis of 24 x 2 mL samples. It is also used for extraction of sensitive biological compounds.

Brand:  MP Biomedicals™ 116002528

1848.53 EUR valid until 2024-12-31
Use promo code "22000" to get your promotional price.


Product Code. 11482360

  • 2295.00 € / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The CoolPrep 24 x 2 mL adapter is a cryogenic adapter which allows simultaneous lysis of 24 x 2 mL samples. Based on passive temperature control technology, the CoolPrep adapter ensures an efficient cooling of the samples as dry-ice, placed into the moving tray is in direct contact with the sample tubes. Due to a high heat transfer capacity and FastPrep™ precise settings of lysis parameters, the samples can be repeatably homogenized in their unthawed state. Most samples can be processed in 40 seconds or less.

  • Application: Ideally suited for extractions of any temperature unstable or sensitive biological compounds such as RNA, siRNA, metabolites, intermediates, and enzymes from even the hardest samples to lyse.
  • Capacity: 20 tubes
  • Sample Volume: Up to 5 g
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cryogenic Adapter
15°C to 30°C
24 x 2 mL
680 g
5 g
Extractions of any temperature-unstable or sensitive biological compounds such as RNA, siRNA, metabolites, intermediates and Enzymes from even the hardest samples to lyse
1.499 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.