missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ FastPrep-24™ Classic Bead Beating Grinder and Lysis System

FastPrep-24™ Classic Bead Beating Grinder and Lysis System allows simultaneous homogenization of up to 24 samples within 40 seconds. It performs a thorough and quick lysis of any tissues and cells. 9 adapters allows a unique versatility in sample number, size and temperature conditions.

Brand:  MP Biomedicals™ 116004500

Product Code. 12079310

  • 10770.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Product Overview: The FastPrep-24™ Classic Instrument is a high-speed benchtop homogenizer offering the ultimate in speed and performance for the lysis of biological samples. Simultaneous homogenization of up to 24 samples takes place within 40 seconds.
  • Service Plan: One-year warranty is included with purchase.
  • Applications: FastPrep™-24 is used to homogenize biological samples. FastPrep™-24 Instrument lyses thoroughly and quickly any tissues and cells and thus allows easy and reproducible isolation of stable RNA, active proteins and full-length genomic DNA.
  • Sample Volume: ∼200 μL to 50 mL; up to ∼25 g
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ideal for extractions of nucleic acids, proteins, metabolites and other small molecules
10.6 in.
50/60 Hz
1200 W
200 μL to 50 mL, up to 25 g
27 cm
90/250 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ FastPrep-24™ Classic Bead Beating Grinder and Lysis System > FastPrep-24™ Classic Bead Beating Grinder and Lysis System

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.