Learn More
MP Biomedicals™ FastRNA™ Pro Green Kit, 2 mL Tubes
FastRNA™ Pro Green Kit is designed to efficiently isolate total RNA from any type of plant and animal tissue or cultured cells.
Brand: MP Biomedicals™ 116045050
329.09 EUR valid until 2024-12-31
Use promo code "21969" to get your promotional price.
Requires : FastPrep Instrument or alternative high speed disruptor, microcentrifuge, pipettmen, RNase Erase, Chloroform, 100% ethanol, 75% ethanol, 1.5 or 2.0 mL RNase-free microcentrifuge tubes, Agarose, Gel loading dye and RNA size marker
This item is not returnable. View return policy
Description
- Product Overview: The FastRNA™ Pro Green Kit is designed to efficiently isolate total RNA from any type of plant and animal tissue or cultured cells. Using the FastPrep-24™ or FastPrep™ FP120 Instrument, between 50 to 500 mg of tissue can be homogenized by Lysing Matrix D in impact-resistant 2 mL tubes. Total RNA is released into the proprietary, protective RNApro™ Solution. Following extraction with chloroform and precipitation, total RNA is extracted.
- Applications: FastRNA™ Pro Green Kit is used to isolate total RNA from any type of plant and animal tissue or cultured cells. Extracted RNA is suitable for RT-PCR analysis and other downstream applications.
- Sample Type: Animal Tissues, Plants, Cultured Cells
- Sample Volume: Up to 500 mg per prep
Specifications
Animal Tissue, Plants, Cultured Cells, Insects, Worms | |
RNApro™ Solution (1 x 55 mL Bottle), DEPC-H2O (1 x 15 mL Bottle), Lysing Matrix D (50 x 2 mL Tubes) | |
Lysing Matrix D | |
50 x 2 mL |
Nucleic Acid isolation and purification, Protein isolation, molecular biology, gene expression, microbiological analysis | |
50 Preps | |
FastRNA-24 Kits |
Safety and Handling
- FastRNA Pro Green Kit
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : phenol, guanidine thiocyanate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.