missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Geneclean™ Turbo Kit for PCR

Geneclean™ Turbo Kit optimized for PCR purification of DNA fragments of sizes 100 bp to 10 kb.

Brand:  MP Biomedicals™ 111103400


 View more versions of this product

Product Code. 11333129

  • 221.00 € / 100 preps

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Ready for direct use
  • Removes primers, nucleotides, enzymes, mineral oil, salts, and other impurities
  • Applications: DNA Clean-up
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Post amplification PCR reaction mixture (oil-free)
GeneClean™ Turbo™ Salt Solution, Wash Solution, Cartridges, Catch Tubes and Elution Solution
Up to 100 μL PCR Reaction Solution
Isolation of DNA from PCR reactions, removal of primers, Nucleotides, Enzymes, Mineral Oil, salts and other impurities from DNA samples
100 Preps
Gels and Solutions
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • geneclean turbo for PCR kits

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : guanidine thiocyanate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.