missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ γ-Globulin from Bovine Plasma

Purity: 98%

Brand:  MP Biomedicals™ 0219147805

 View more versions of this product

Product Code. 11484702

  • 154.76 € / 5g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

White, crystalline, lyophilized powder.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

25mg/mL
The product is lyophilized, in normal Saline solution (25mg/mL, 0.9%NaCl)
g-Globulin from Bovine Plasma
5 g
0°C
Cell Biology and Analysis, Genomics, Molecular Biology
∽150000
6 to 8
Bovine Plasma
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers