missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ γ-Globulins, Bovine Plasma, 100g

99.3%, Lyophilized

Brand:  MP Biomedicals™ 08820414

Product Code. 11474246

  • 2545.00 € / 100g
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

White to Off-white
6.7
2°C to 8°C
Used as a co-precipitant in RIA methods employing Polyethylene Glycol or Ammonium Sulfate precipitate
Bovine Plasma
100%
100 g
Lyophilized
99.3%
Bovine Plasma gamma Globulin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers