missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Glucose Oxidase

Product Code. p-3563480 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 kU
250 kU
50 kU
500 kU
Unit Size:
10ku
250ku
500ku
50ku
This item is not returnable. View return policy

Product Code. 11481092

missing translation for 'mfr': MP Biomedicals™ 0219519610

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

From Aspergillus niger

Type X. Activity: >100U/mg. Unit Definition: one unit oxidizes 1.0μmol of β-D-glucose to D-gluconic acid and H2O2 per minute at 35°C, pH 5.1.

TRUSTED_SUSTAINABILITY

Especificaciones

Concentration >100 U/mg
Color Yellow
pH 4 to 7
For Use With (Application) Glucose Oxidase can be utilized in the enzymatic determination of D-glucose in solution
Content And Storage -20°C
Source Aspergillus niger
Quantity 10 kU
Product Type Glucose Oxidase
Molecular Weight (g/mol) ≈160000
Inhibitors Ag+, Hg2+, Cu2+ , 4-chloromercuribenzoate, 50% D-arabinose
Form Lyophilized Powder
Mostrar más Mostrar menos
compliance-icons
Palabra de advertencia
  • Danger
Clasificación
  • Respiratory sensitiser Category 1
Indicaciones de peligro
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Consejos de prudencia
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Otros peligros
  • MIXTURE LIST-Contains : glucose oxidase
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.