Learn More
MP Biomedicals™ 1X Hanks' Balanced Salt Solution (Modified) without Phenol Red
1X Hanks' Balanced Salt Solution (Modified) without Phenol Red is a clear medium ideal for working with stem cells or when growing cells at low densities.
Brand: MP Biomedicals™ 091810254
30.43 EUR valid until 2024-12-31
Use promo code "21951" to get your promotional price.
This item is not returnable.
View return policy
Description
- Phenol red has been shown to interfere with the growth of some cells at cloning densities.
- Use this medium when working with stem cells or when growing cells at low densities.
Specifications
Clear | |
1 X | |
500 mL | |
6.9 to 7.4 |
15°to 30°C | |
Hanks' Balanced Salt Solution (HBSS) | |
Calcium Chloride 140 mg/L, Magnesium Sulfate 97.7 mg/L, Potassium Chloride 400 mg/L, Potassium Phosphate Monobasic 60 mg/L, Sodium Bicarbonate 350 mg/L, Sodium Chloride 8000 mg/L, Sodium Phosphate Dibasic 47.7 mg/L, Dextrose 1000 mg/L | |
Liquid |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.