missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lysing Matrix F, 2 mL Tubes

Lysing Matrix F, 2 mL Tubes are used to isolate RNA and protein from samples. Lysing Matrix F is useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing.

Brand:  MP Biomedicals™ 116915500

1487.71 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15242830

  • 1880.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Impact-resistant 2 mL tubes contain 1.6 mm aluminum oxide particles and 1.6 mm silicon carbide particles
  • With white caps
  • Specific Gravity: Aluminum Oxide, 3.95; Silicon Carbide, 3.20
  • Nucleic Acid Isolation and Purification
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2 mL
Dnase-free
Lysing matrix F
500 x 2 mL Tubes
Bacteria (Gram +/-), Animal Tissues, Fungi, Plants, Coral Emulsions, Mold
Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis
Aluminum Oxide, Al2O3; Silicon Carbide, SiC
RNase-free
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.