missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lysing Matrix H

Lysing Matrix H used to isolate RNA and Proteins from bacterial aggregates, biofilms, and whole insects.

Brand:  MP Biomedicals™ 116917050

 View more versions of this product

Product Code. 11422430

  • 227.00 € / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Impact-resistant 2 mL tube contains 2 mm glass beads and 2 mm yellow zirconium oxide beads
  • With yellow caps
  • Specific Gravity: Glass Beads, 2 mm; Yellow Zirconium Oxide Beads, 5.5
  • Applications: Nucleic Acid Isolation and Purification
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Glass; Yellow Zirconium Oxide
Seeds, Animal Tissues, Plants, Soil, Bacteria, Ancient and Dried Samples, Clay, Wood
Nucleic Acid isolation and purification
50 x 2 mL Tubes
2 mL
Dnase-free
Lysing matrix H
RNase-free
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.