missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lysing Matrix J
Lysing Matrix J used for nucleic acid isolation and purification.
Brand: MP Biomedicals™ 116919500
This item is not returnable.
View return policy
Description
- Average size: Yellow zirconium oxide beads, Vickers 900 to 1000, aluminum oxide, Knoop 2000 and Mohs 9
- Material construction: Matrix J, contains 2 mm yellow zirconia beads and 1.6 mm aluminum oxide granules
- Sample type: Plants, animal tissue, bacteria (gram +/-), mold, fungi, coral emulsions and fixed samples
- Instrument compatibility: FastPrep-24, FastPrep 120, FastPrep-96
- Specific gravity: Yellow zirconium oxide beads, 5.5; Aluminum oxide 3.95
- Tube compatibility: 2 mL
- Matrix type: J

Specifications
Yellow Zirconium Oxide, Aluminum Oxide | |
Bacteria (Gram +/-), Animal Tissues, Fungi, Plants, Fixed Samples, Coral Emulsions, Mold | |
Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis | |
500 x 2 mL Tubes |
2 mL | |
Dnase-free | |
Lysing matrix J | |
Rnase-free |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Lysing Matrix J > Lysing Matrix J; Quantity: 500 x 2 mL Tubes
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction