missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lysozyme from Chicken Egg White, 20,000U/mg
Hydrolyzes b-1,4 glucosidic linkages between N-acetylmuramic acid and N-acetylglucosamine which occur in mucopeptide cell wall structure of certain microorganisms, such as Micrococcus lysodeikticus
118.00 € - 1635.00 €
Specifications
Concentration | 3X |
---|---|
Color | White |
pH | 11 |
For Use With (Application) | Lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific Antigen |
Content And Storage | 0°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11394019
|
MP Biomedicals™
0210083105 |
5 g |
118.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304029
|
MP Biomedicals™
0210083110 |
10 g |
233.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314029
|
MP Biomedicals™
0210083125 |
25 g |
408.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324029
|
MP Biomedicals™
0210083180 |
100 g |
1635.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Synonyms: Muramidase, Mucopeptide N-acetylmuramoylhydrolase
- CAS Number: 9001-63-2
- EC Number: 232-620-4
- MDL Number: MFCD00131557
- Format: 3X crystallized, salt-Free, albumin-Free, lyophilized
- Specific Activity: ≤20,000U/mg solid
- Extinction Coefficient (E1%): 26.4 (280 nm)
- Isoelectric point (pI): pH 11.0
- pH: 3.6
Used for lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific antigen. Also acts as germinative agent of bacterial spores. In humans, lysozyme may mediate anti-tumor function of macrophages. Cartilage lysozyme has a role in cartilage calcification.
Specifications
3X | |
11 | |
0°C | |
Lysozyme | |
Surface-active reagents such as Dodecyl Sulfate, Alcohols and Fatty Acids, Imidazole and Indole derivatives are inhibitors via formation of change-transfer complexes |
White | |
Lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific Antigen | |
Chicken egg white | |
14388 | |
Crystallized, Lyophilized |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- lysozyme
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : lysozyme