Learn More
MP Biomedicals™ Mouse Diet, Purified, Pellet
Natural-ingredient diet specifically formulated to provide the proper balance of all known nutrients considered essential for the growth, maintenance, and reproduction of mice for lab experiment
Brand:  MP Biomedicals™ 0290460610
 View more versions of this product
Product Code. 11465732
  • 875.00 € / 10kg
Explore more special offers
Description
Show More
Specifications
Specifications
Animal Diet
Pellet
10 kg
15°C to 30°C
Vitamin Free Casein 30%, Sucrose 52.8%, Alphacel, Non-nutritive Bulk 2%, Corn Oil 10%, Cod Liver Oil, USP 0.2%, Salt Mixture Sure No. 2 5%
Mouse diet is natural ingredient diet specifically formulated to provide the proper balance of all known nutrients considered essential for the growth, maintenance and reproduction of Mice for lab experiment
Mouse
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals™ Mouse Diet, Purified, Pellet > Mouse Diet, Purified, Pelleted, 10 kg