missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Mycoplasma Removal Agent (MRA)
Mycoplasma Removal Agent (MRA) is a derivative of the quinoline family of antibiotics and eliminates mycoplasma contamination in cell cultures by inhibiting mycoplasma DNA gyrase.
Brand: MP Biomedicals™ 093050044
411.01 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Derivative of the quinoline family of antibiotics and eliminates mycoplasma infection by inhibiting mycoplasma DNA gyrase
- Supplied as a ready-to-use solution
- Cited by the European Collection of Animal Cell Cultures (Porton Down, U.K.) as being superior to all other current products for the eradication of mycoplasma
- Cell toxicity is low following treatment at the recommended concentration of 0.5 μg/mL
- Eliminates multiple mycoplasma species within one week using the lowest dosage
- Prevents recontamination of the culture at 0.1 μg/mL
- Maintains cell viability
- Compatible with most mammalian cell lines
- Sterility and low cytotoxicity
Specifications
Store at Room Temperature (15 to 30°C). Protect from light. | |
Clear, light yellow solution | |
Sterile | |
5 mL |
50 µg/mL | |
Mycoplasma Removal | |
Mycoplasma elimination |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction