Learn More
MP Biomedicals™ Cover for PlantCon™ Plant Tissue Culture Container
Plastic cover for PlantCon™ Plant Tissue Culture Container. Use PlantCon™ containers with liquid or agar solidified media. They can be used for many plant cell culture procedures that currently utilize glass containers.
Brand: MP Biomedicals™ 092672102
51.90 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 4.08230kg
Description
- Cover can be snapped on tight, for long-term culture, or loosely fitted during acclimation to external conditions prior to transplantation
- Ideal for use in commercial horticulture or research settings
- Allow exceptional light transmission and optimal growth
- Reduce the time spent preparing an in vitro growth environment for plant cultures by eliminating the need for cleaning and sterilization
Specifications
Plastic | |
Clear | |
PVC | |
Sterile | |
Optimal Plant Cell Culture Applications | |
10/Cs | |
3.75 x 3.75 in. (9.5 x 9.5 cm) | |
9.5 cm | |
7 cm |
Square | |
Single-use | |
Rectangular | |
Cell Culture Dish Cover | |
PlantCon™ Containers (092672206, 092672002) | |
Yes | |
9.5 x 9.5 x 7 cm | |
9.5 cm |