missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Poly-L-Lysine Hydrobromide Attachment Factors: ≥70,000 MW
Facilitate attachment, adherence, and spreading in the culture of adherent cells
Brand: MP Biomedicals™ 0215017780
275.46 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Available nonsterile or gamma-irradiated.
Specifications
-20°C | |
Non-sterile | |
Synthetic |
Poly-L-Lysine | |
100 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction