missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Rat Serum
Sterile, filtered, Lyophilized Serum
Brand: MP Biomedicals™ 08642941
This item is not returnable.
View return policy
Description
Specifications
-20°C | |
Lyophilized | |
10 mL | |
Sterile | |
Blocking agents and controls in immunoassays | |
Yes |
Rat | |
Rat Serum | |
5.5 to 7 g/dL | |
Filtration | |
2.34 mg/dL | |
7.8 |